![2-Bromo-4-ethylpyridine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/316/566/07e02e26-9388-4c9b-8ebb-8ac0a885be69/640/07e02e26-9388-4c9b-8ebb-8ac0a885be69.png)
2-Bromo-4-ethylpyridine
![2-Bromo-4-ethylpyridine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/316/566/07e02e26-9388-4c9b-8ebb-8ac0a885be69/640/07e02e26-9388-4c9b-8ebb-8ac0a885be69.png)
Synonym(s):
4-Ethyl-2-bromopyridine
Empirical Formula (Hill Notation):
C7H8BrN
CAS Number:
Molecular Weight:
186.05
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing