Skip to Content
Merck
2-Amino-6-chloropurine ≥99%

2-Amino-6-chloropurine

2-Amino-6-chloropurine ≥99%
Synonym(s):
6-Chloro-2-purinamine, 6-Chloroguanine
Empirical Formula (Hill Notation):
C5H4ClN5
CAS Number:
Molecular Weight:
169.57
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing