![2,4,7,9-Tetramethyl-5-decyne-4,7-diol ethoxylate average Mn 670](/deepweb/assets/sigmaaldrich/product/structures/204/325/c2217fad-0e2a-497b-a360-5872d7bafd42/640/c2217fad-0e2a-497b-a360-5872d7bafd42.png)
2,4,7,9-Tetramethyl-5-decyne-4,7-diol ethoxylate
![2,4,7,9-Tetramethyl-5-decyne-4,7-diol ethoxylate average Mn 670](/deepweb/assets/sigmaaldrich/product/structures/204/325/c2217fad-0e2a-497b-a360-5872d7bafd42/640/c2217fad-0e2a-497b-a360-5872d7bafd42.png)
Linear Formula:
(CH3)2CHCH2C(CH3)[(-OCH2CH2-)mOH]C≡CC(CH3)[(-OCH2CH2-)nOH]CH2CH(CH3)2
CAS Number:
Products
Brand
Boiling Point (°C)
pH Value
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing