2,3-Dihydroxypyridine
Synonym(s):
2,3-Pyridinediol
Empirical Formula (Hill Notation):
C5H5NO2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing