Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-(2-Aminoethyl)isothiourea dihydrobromide
Synonym(s):
2-(2-Aminoethyl)-2-thiopseudourea dihydrobromide, AET, S-(2-Aminoethyl)isothiouronium bromide hydrobromide
Linear Formula:
NH
2
CH
2
CH
2
SC(=NH)NH
2
· 2HBr
CAS Number:
56-10-0
Molecular Weight:
281.01
EC Number:
200-257-0
Products
Product Category
bioactive small molecules (1)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Research Area
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
A5879
2-(2-Aminoethyl)isothiourea dihydrobromide,
Expand
View Pricing