Skip to Content
Merck
2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate mixture of isomers, 99%

2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate

2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate mixture of isomers, 99%
Linear Formula:
(CH3)2CHCH[O(R)]C(CH3)2CH2O(R), R=-COCH(CH3)2 or H
CAS Number:
Molecular Weight:
216.32
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing