Skip to Content
Merck
2-[2-[4-(Diphenylmethyl)-1-piperazinyl]ethoxy]ethanol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-[2-[4-(Diphenylmethyl)-1-piperazinyl]ethoxy]ethanol

2-[2-[4-(Diphenylmethyl)-1-piperazinyl]ethoxy]ethanol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Decloxizine
Empirical Formula (Hill Notation):
C21H28N2O2
CAS Number:
Molecular Weight:
340.46

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade