(1R,2R)-(+)-1,2-Diphenylethylenediamine
Synonym(s):
(1R,2R)-(+)-1,2-Diamino-1,2-diphenylethane
Linear Formula:
[C6H5CH(NH2)-]2
CAS Number:
Molecular Weight:
212.29
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing