Skip to Content
Merck
1-Ethyl-3-methylimidazolium trifluoromethanesulfonate for electrochemistry, ≥98.0% (T)

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate for electrochemistry, ≥98.0% (T)
Synonym(s):
EMIM Otf
Empirical Formula (Hill Notation):
C7H11F3N2O3S
CAS Number:
Molecular Weight:
260.23

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade