Skip to Content
Merck
1-Bromo-2-methyl-1-propene 98%

1-Bromo-2-methyl-1-propene

1-Bromo-2-methyl-1-propene 98%
Synonym(s):
Isocrotyl bromide
Linear Formula:
(CH3)2C=CHBr
CAS Number:
Molecular Weight:
135.00

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing