Skip to Content
Merck
vibrant-m

(16α)-21-(Acetyloxy)-17-hydroxy-16-methylpregna-1,4,9(11)-triene-3,20-dione

vibrant-m
Synonym(s):
17-Hydroxy-16α-methyl-3,20-dioxopregna-1,4,9(11)-trien-21-yl acetate
Empirical Formula (Hill Notation):
C24H30O5
CAS Number:
Molecular Weight:
398.49

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Special Grade