Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,5-Diazabicyclo[4.3.0]non-5-ene
Synonym(s):
DBN, 1,5-Diazabicyclo[4.3.0]non-5-ene
Empirical Formula (Hill Notation):
C
7
H
12
N
2
CAS Number:
3001-72-7
Molecular Weight:
124.18
EC Number:
221-087-3
Products
Product Category
organic bases (2)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
136581
1,5-Diazabicyclo[4.3.0]non-5-ene,
98%
Expand
View Pricing
8.03603
1,5-Diazabicyclo[4.3.0]non-5-ene,
for synthesis
Expand
View Pricing
33471
1,5-Diazabicyclo[4.3.0]non-5-ene,
purum, ≥98.0% (GC)
Expand
View Pricing