![1,4-Butane sultone ≥99%](/deepweb/assets/sigmaaldrich/product/structures/202/289/1443c8d6-ce73-4813-bdd8-89fe620eb79f/640/1443c8d6-ce73-4813-bdd8-89fe620eb79f.png)
1,4-Butane sultone
![1,4-Butane sultone ≥99%](/deepweb/assets/sigmaaldrich/product/structures/202/289/1443c8d6-ce73-4813-bdd8-89fe620eb79f/640/1443c8d6-ce73-4813-bdd8-89fe620eb79f.png)
Synonym(s):
4-Hydroxybutane-1-sulfonic acid δ-sultone, 1,4-Butanesultone
Empirical Formula (Hill Notation):
C4H8O3S
CAS Number:
Molecular Weight:
136.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing