Skip to Content
Merck
1,3,5-Tris(2-hydroxyethyl)isocyanurate 97%

1,3,5-Tris(2-hydroxyethyl)isocyanurate

1,3,5-Tris(2-hydroxyethyl)isocyanurate 97%
Synonym(s):
1,3,5-Tris(2-hydroxyethyl)-1,3,5-triazine-2,4,6(1H,3H,5H)-trione
Empirical Formula (Hill Notation):
C9H15N3O6
CAS Number:
Molecular Weight:
261.23
EC Number:

Products

Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing