Skip to Content
Merck
12-Hydroxystearic acid United States Pharmacopeia (USP) Reference Standard

12-Hydroxystearic Acid

12-Hydroxystearic acid United States Pharmacopeia (USP) Reference Standard
Synonym(s):
12-Hydroxyoctadecanoic acid, 12-HSA, 12-Hydroxystearic acid
Linear Formula:
CH3(CH2)5CH(OH)(CH2)10COOH
CAS Number:
Molecular Weight:
300.48

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing