Skip to Content
Merck
1-(2,3-Dichlorophenyl)piperazine hydrochloride 97%

1-(2,3-Dichlorophenyl)piperazine hydrochloride

1-(2,3-Dichlorophenyl)piperazine hydrochloride 97%
Synonym(s):
1-(2,3-dichlorophenyl)piperazine, 2,3-DCPP HCl
Linear Formula:
C10H12Cl2N2 · HCl
CAS Number:
Molecular Weight:
267.58

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing