Skip to Content
Merck
11-Mercapto-1-undecanol 99%

11-Mercapto-1-undecanol

11-Mercapto-1-undecanol 99%
Synonym(s):
11-Hydroxy-1-undecanethiol, 11-Hydroxy-1-undecanthiol, 11-Hydroxyundecane-1-thiol, 11-Mercaptoundecanol, MUD
Linear Formula:
HS(CH2)11OH
CAS Number:
Molecular Weight:
204.37

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing