Skip to Content
Merck
1,1-Diphenyl-2-propyn-1-ol 99%

1,1-Diphenyl-2-propyn-1-ol

1,1-Diphenyl-2-propyn-1-ol 99%
Linear Formula:
HC≡CC(C6H5)2OH
CAS Number:
Molecular Weight:
208.26

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing