Skip to Content
Merck
All Photos(1)

Documents

V900917

Isopropyl β-D-thiogalactoside

Vetec, reagent grade, ≥99%

Synonym(s):

Isopropyl β-D-1-thiogalactopyranoside, IPTG, Isopropyl β-D-thiogalactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O5S
CAS Number:
Molecular Weight:
238.30
Beilstein:
4631
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥99%

storage temp.

2-8°C

SMILES string

CC(C)S[C@@H]1OC(CO)[C@H](O)C(O)[C@H]1O

InChI

1S/C9H18O5S/c1-4(2)15-9-8(13)7(12)6(11)5(3-10)14-9/h4-13H,3H2,1-2H3/t5-,6+,7+,8-,9+/m1/s1

InChI key

BPHPUYQFMNQIOC-NXRLNHOXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

IPTG is commonly used in cloning procedures that require induction of β-galactosidase activity. It is used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies that induce expression of the lac operon in Escherichia coli. IPTG functions by binding to the lacI repressor and altering its conformation, which prevents the repression of the β-galactosidase coding gene lacZ.

Other Notes

Non-metabolizable galactose analog.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Therese Wohlschlager et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), E2787-E2796 (2014-06-01)
Effector proteins of innate immune systems recognize specific non-self epitopes. Tectonins are a family of β-propeller lectins conserved from bacteria to mammals that have been shown to bind bacterial lipopolysaccharide (LPS). We present experimental evidence that two Tectonins of fungal
Farshid Hadifar et al.
PloS one, 9(10), e108420-e108420 (2014-10-21)
Killed avian influenza virus (AIV) vaccines have been used to control H5N1 infections in countries where the virus is endemic. Distinguishing vaccinated from naturally infected birds (DIVA) in such situations however, has become a major challenge. Recently, we introduced the
Iltaf Ahmed et al.
Human molecular genetics, 24(11), 3172-3180 (2015-02-24)
There are two known mRNA degradation pathways, 3' to 5' and 5' to 3'. We identified likely pathogenic variants in two genes involved in these two pathways in individuals with intellectual disability. In a large family with multiple branches, we
Laam Li et al.
BMC genomics, 16, 637-637 (2015-08-27)
Legionella pneumophila (Lp) is a water-borne opportunistic pathogen. In water, Lp can survive for an extended period of time until it encounters a permissive host. Therefore, identifying genes that are required for survival in water may help develop strategies to
Tia DiTommaso et al.
PLoS genetics, 10(10), e1004706-e1004706 (2014-10-24)
Keratins are cytoskeletal intermediate filament proteins that are increasingly being recognised for their diverse cellular functions. Here we report the consequences of germ line inactivation of Keratin 76 (Krt76) in mice. Homozygous disruption of this epidermally expressed gene causes neonatal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service