Skip to Content
Merck
All Photos(2)

Key Documents

V900401

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide hydrate

Vetec, reagent grade, ≥96.5%

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

≥96.5%

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2.O[C@@H]3[C@@H](COP(O)(OP(OC[C@@H](O4)[C@@H](O)[C@@H](O)[C@H]4[N+]5=CC=CC(C(N)=O)=C5)([O-])=O)=O)OC[C@@H]3O

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1

InChI key

BAWFJGJZGIEFAR-NNYOXOHSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Nicotinamide adenine dinucleotide (NAD) is a ubiquitously[1] found electron carrier[2] and a cofactor. NAD+ contains an adenylic acid and a nicotinamide-5′-ribonucleotide group linked together by a pyrophosphate moiety. In NAD+ complexes, the enzyme-cofactor interactions are highly conserved.[1]

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide (NAD) significantly participates in enzyme-catalyzed oxido-reduction processes[1] and many genetic processes.[3] It cycles between the oxidized (NAD+) and reduced (NADH) forms to maintain a redox balance necessary for continued cell growth. NAD is also involved in microbial catabolism.[3]
Electron acceptor

Packaging

Packaged by solid weight.

Other Notes

This is the common form of NAD.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental and theoretical electron density studies in large molecules: NAD+, beta-nicotinamide adenine dinucleotide
Guillot B, et al.
The Journal of Physical Chemistry B, 107(34), 9109-9121 (2003)
A Mechanism of Adsorption of beta-Nicotinamide Adenine Dinucleotide on Graphene Sheets: Experiment and Theory
Pumera M, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 15(41), 10851-10856 (2009)
Metabolic engineering of Escherichia coli: increase of NADH availability by overexpressing an NAD+-dependent formate dehydrogenase.
Berrios-Rivera S J, et al.
Metabolic engineering, 4(3), 217-229 (2002)
Ana P Gomes et al.
Cell, 155(7), 1624-1638 (2013-12-24)
Ever since eukaryotes subsumed the bacterial ancestor of mitochondria, the nuclear and mitochondrial genomes have had to closely coordinate their activities, as each encode different subunits of the oxidative phosphorylation (OXPHOS) system. Mitochondrial dysfunction is a hallmark of aging, but
Christian Dölle et al.
The FEBS journal, 280(15), 3530-3541 (2013-04-27)
Mitochondrial metabolism is intimately connected to the universal coenzyme NAD. In addition to its role in redox reactions of energy transduction, NAD serves as substrate in regulatory reactions that lead to its degradation. Importantly, all types of the known NAD-consuming

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service