Skip to Content
Merck
All Photos(1)

Documents

SY3H95ECD606

Synthonix Corporation

7-Bromo-4-chloroimidazo[2,1-f][1,2,4]triazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
233.45
MDL number:

Iupac Name

7-bromo-4-chloroimidazo[2,1-f][1,2,4]triazine

Inchi Code

1S/C5H2BrClN4/c6-3-1-8-5-4(7)9-2-10-11(3)5/h1-2H

InChI key

RRXLWAFFBIFFKO-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/11600.html

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service