Skip to Content
Merck
All Photos(1)

Key Documents

Y0002405

Sotalol for system suitability CRS

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. Compendial materials should be used for the purpose described in the appropriate monograph unless otherwise authorized by the National Competent Authority. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

e.g. pharmaceutical (small molecule)

Packaging

Unit quantity: 10mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service