Skip to Content
Merck
All Photos(1)

Documents

G001056

HPLC Analysis of Barbiturates in Serum on Discovery® C18 after SPE using Discovery® DSC-18Lt

application for SPE, application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable
solid phase extraction (SPE): suitable

test parameters

sample preparation: SPE (Solid Phase Extraction)
sample/matrix: 0.5 mL porcine serum spiked with 0.5 μg/mL or 1.0 μg/mL each analyte then diluted with 0.5 mL water
SPE tube/cartridge: Discovery DSC-18Lt, 500 mg/3 mL (52613-U)
condition: 2 mL methanol; 2 mL DI water
sample addition: 1 mL at 0.75 mL/min
washing: 2 mL 5% methanol, then vacuum or air dry for 5-10 min
elution: 1-2 mL methanol
eluate post-treatment: dry eluate with nitrogen purge (40 °C; 15-20 min), reconstitute in 20 μL mobile phase
column: Discovery C18, 15 cm × 4.6 mm, 5 μm preceded by a 2 cm C18 guard column and 0.5 μm frit filter (504955)
mobile phase: [A] methanol; [B] water (40:60, A:B)
flow rate: 1 mL/min
column temp.: 30 °C
detector: UV, 214 nm
injection: 30 μL, diluted porcine serum extract

suitability

application for HPLC
application for SPE

application(s)

forensics and toxicology

Analysis Note

Barbiturates are commonly abused and among the most widely tested compounds in clinical, forensic, or therapeutic drug monitoring applications. Shown here is the baseline separation of a set of barbiturates on a Discovery C18 HPLC column after extraction from spikd serum with Discovery DSC-18Lt SPE. Highest grade HPLC solvents were used to supply low background interference and low particulate contamination for robust, trouble-free operation. Cerilliant and Sigma-Aldrich reference standards provided reliable identification and quantification. A Zymark® RapidTrace® SPE Workstation was used in this study.

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany
RapidTrace is a registered trademark of Zymark Corp.
Zymark is a registered trademark of Sotax Corporation

Analyte

Description

    Phenobarbital solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Aprobarbital
    Butabarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Mephobarbital, United States Pharmacopeia (USP) Reference Standard
    Pentobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Secobarbital solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service