Skip to Content
Merck
All Photos(1)

Documents

40091

Supelco

4,4′-DDE solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene solution, 4,4′-DDE solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4)2C=CCl2
CAS Number:
Molecular Weight:
318.03
Beilstein:
1913355
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

room temp

SMILES string

Clc1ccc(cc1)\C(=C(/Cl)Cl)c2ccc(Cl)cc2

InChI

1S/C14H8Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8H

InChI key

UCNVFOCBFJOQAL-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mireia Gascon et al.
Environment international, 52, 51-65 (2013-01-08)
Disruption of developing immune and respiratory systems by early-life exposure to persistent organic pollutants (POPs) could result into reduced capacity to fight infections and increased risk to develop allergic manifestations later in life. To systematically review the epidemiologic literature on
Luisa Torres-Sánchez et al.
Environmental health perspectives, 121(2), 263-268 (2012-11-16)
The results of previous studies suggest that prenatal exposure to bis[p-chlorophenyl]-1,1,1-trichloroethane (DDT) and to its main metabolite, 2,2-bis(p-chlorophenyl)-1,1-dichloroethylene (DDE), impairs psychomotor development during the first year of life. However, information about the persistence of this association at later ages is
Loretta A Fernandez et al.
Environmental science & technology, 46(21), 11937-11947 (2012-10-16)
Passive sampling was used to deduce water concentrations of persistent organic pollutants (POPs) in the vicinity of a marine Superfund site on the Palos Verdes Shelf, California, USA. Precalibrated solid phase microextraction (SPME) fibers and polyethylene (PE) strips that were
Anita H Poulsen et al.
Environmental pollution (Barking, Essex : 1987), 175, 92-99 (2013-01-29)
This study evaluated the dietary uptake kinetics and sublethal toxicity of p,p'-dichlorodiphenyl dichloroethylene (p,p'-DDE) in Antarctic krill. The uptake rate constant (characterised by the seawater volume stripped of contaminant sorbed to algae) of 200 ± 0.32 mL g(-1) wet weight
Roberto De La Torre-Roche et al.
Environmental science & technology, 47(2), 718-725 (2012-12-21)
The effect of nanoparticle (NP), bulk, or ionic Ag exposure on dichlorodiphenyldichloroethylene (p,p'-DDE; DDT metabolite) accumulation by Glycine max L. (soybean) and Cucurbita pepo L. (zucchini) was investigated. The plants were grown in 125-mL jars of vermiculite amended with 500

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service