Skip to Content
Merck
All Photos(1)

Documents

30988

Supelco

Flutolanil

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N-(3-Isopropoxyphenyl)-2-(trifluoromethyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16F3NO2
CAS Number:
Molecular Weight:
323.31
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(OC(C)C)=CC=C1)C2=CC=CC=C2C(F)(F)F

InChI

1S/C17H16F3NO2/c1-11(2)23-13-7-5-6-12(10-13)21-16(22)14-8-3-4-9-15(14)17(18,19)20/h3-11H,1-2H3,(H,21,22)

InChI key

PTCGDEVVHUXTMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Flutolanil is a systemic amide fungicide with curative and protective action, belonging to the chemical class of phenylbenzamide. It is widely used in crops, fruits, vegetables, and some ornamentals to control diseases caused by basidiomycetes. It is effective against rice sheath blight, wheat sharp eyespot, potato stem canker, black scurf, peanut stem rot, tall fescue brown patch caused by Rhizoctonia, and other plant diseases caused by Corticium, Typhula, and Gymnosporangium. Flutolanil acts by inhibiting the synthesis of succinate dehydrogenase (SDH), in mitochondrial complex II, disrupting the Krebs cycle and the mitochondrial electron transport chain.

According to Regulation (EC) 1107/2009 (repealing 91/414/EEC) flutolanil is approved for use in European Union (EU). The maximum residue level (MRL) for flutolanil, following Reg. (EU) 2021/2202, is set from 0.01 mg/kg to 2 mg/kg for products of plant and animal origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The flutolanil CRM may also find following uses:
  • Examine the reproductive endpoints on the gonad endocrine system of the parental zebrafish after chronic exposure to different concentrations of flutolanil for 60 days
  • Spectroscopic analysis of acetamiprid, flutolanil, and etofenprox residues extracted from tomato fruits
  • Development and validation of two robust chromatographic methods for estimation of tomatoes specific acetamiprid, flutolanil, and etofenprox residues on fruits before food processing and serving
  • To investigate the developmental toxicity and the potential mechanism of thyroid endocrine disruption induced by flutolanil in zebrafish embryos based on 1H NMR metabolomics analysis
  • To develop a sensitive LC-MS/MS analytical method for the determination of residual flutolanil and its principal metabolites and conjugates, in livestock and seafood products

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service