Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29842-U

eVol Electronic syringe

pkg of 1 ea

Synonym(s):

eVOL® Components

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

Pricing and availability is not currently available.

feature

programmable dispensing system

packaging

pkg of 1 ea

manufacturer/tradename

SGE 2910005

L × W

8 in. × 1.5 in.

Looking for similar products? Visit Product Comparison Guide

General description

Touch Wheel Control
A full-color display and a convenient touch wheel controller make using the eVol easy. The touch wheel uses a menu-driven approach similiar to popular music devices. Intuitive functions include help screeens and prompts make programming and use effortless.

Other Notes

Limited Warranty:
eVol is warranted by SGE to meet stated quality and performance specifications and to be free of defects in material or workmanship for a period of 1 (one) year from the date of purchase.

Register Warranty: Please fill out the warrant registration on the SGE eVol website www.sge.com/eVol.

Legal Information

eVol is a registered trademark of SGE Analytical Science Pty Ltd.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service