Skip to Content
Merck
All Photos(10)

Documents

ZRB2464

Sigma-Aldrich

Anti-CD40 Antibody, clone 2E20 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

B-cell surface antigen CD40, Bp50, CD40L receptor, CDw40, Tumor necrosis factor receptor superfamily member 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E20, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 30.62 kDa
observed mol wt ~38 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

ELISA: suitable
affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... CD40(958)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2E20 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects CD40. It targets an epitope within 20 amino acids within the extracellular domain from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal half of human CD40.

Application

Quality Control Testing

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD40 in Raji cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CD40 in lysates from THP-1 cells treated with LPS and Human tonsil tissue, and with His/ABP-tagged recombinant fragment of human CD40

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CD40 in Human tonsil tissue sections.

Enzyme Immunoassay (ELISA): A serial of dilutions from a representative lot detected His/ABP-tagged recombinant fragment of human CD40

Affinity Binding Assay: A representative lot of this antibody bound CD40 peptide with a KD of 7.6 x 10-9 in an affinity binding assay.

Flow Cytometry Analysis: 1 µg from a representative lot detected CD40 in one million Human peripheral blood mononuclear cells (PBMC) co-stained with CD19APC.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Tumor necrosis factor receptor superfamily member 5 (UniProt: P25942; also known as B-cell surface antigen CD40, Bp50, CD40L receptor, CDw40, CD40) is encoded by the CD40 (also known as TNFRSF5) gene (Gene ID: 958) in human. CD40, a member of the TNFR super family, is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-20 ), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 21-193), a transmembrane domain (aa 194-215), and a cytoplasmic domain (aa 216-277). It has multiple cysteine-rich repeats. CD40 is expressed in B-cells, monocytes, dendritic cells, thymic epithelium as well as on certain carcinomas. It serves as a receptor for CD40L that is transiently expressed on T-cells and some non-immune cells under inflammatory conditions. Upon engagement with its ligand, CD40 directly or indirectly recruits TRAF1, TRAF2, TRAF3, TRAF5, and TRAF6 to its cytoplasmic domains. The canonical consensus binding sites for TRAF1, TRAF2, and TRAF3 are situated at the membrane distal domain of the intracellular tail of CD40 and is defined by the amino acid sequence PxQxT. CD40 transduces TRAF6 and MAP3K8-mediated signals that activate ERK in macrophages and B cells, leading to induction of immunoglobulin secretion. Clone G28.5 is reported to bind to an epitope localized in D1/B2 domains in the extracellular cysteine rich region and is a potent activator of B-cells. Mutations in CD40 gene are known to cause immunodeficiency with hyper-IgM3 (HIGM3) that is characterized by normal or elevated serum IgM levels with absence of IgG, IgA, and IgE. This results in a profound susceptibility to bacterial infections. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Elgueta, R., et al. (2009). Immunol. Rev. 229(1); 152-172; Malmborg Hager, AC., et al. (2003). Scand. J, Immunol. 57(6); 517-524).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service