Skip to Content
Merck
All Photos(11)

Documents

ZRB2343

Sigma-Aldrich

Anti-β Galactosidase Antibody, clone 2C15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:3.2.1.23;Acid beta-galactosidase;Lactase, Elastin receptor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C15, recombinant monoclonal

description

2C15 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 76.08 kDa
observed mol wt ~120 kDa

purified by

using Protein A

species reactivity

human, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... GLB1(2720)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2C15 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects β-Galactosidase. It targets an epitope within 22 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the N-terminal region of human β-Galactosidase.

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysates.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected β Galactosidase in HepG2 cell lysates.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected β Galactosidase in COS-7 cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound Beta Galactosidase peptide with a KD of 3.5 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected β Galactosidase in human liver tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected β Galactosidase in HepG2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Beta-galactosidase (UniProt: P16278; also known as EC:3.2.1.23, Acid beta-galactosidase, Lactase, Elastin receptor 1) is encoded by the GLB1 (also known as ELNR1) gene (Gene ID: 2720) in human. The β-galactosidase is a highly glycosylated, homodimeric enzyme that cleaves β-linked terminal galactosyl residues from gangliosides, glycoproteins, and glycosaminoglycans. It is also reported to form a tetrameric structure. It is abundantly expressed in placenta and in testis. It displays strict specificity at the galactosyl position but can hydrolyze β-D-galactopyranosides with a wide variety of aglycons. Human β-galactosidase is synthesized with a 23 amino acid secretion signal and the enzyme is synthesized as an 88 kDa precursor that is transferred to the lysosomal compartment where it is processed to a 64 kDa mature enzyme by proteolytic cleavage of the C-terminal region. The C-terminal fragment is reported to remain associated with the N-terminal fragment. In human β-galactosidase, glutamic acid 268 is identified as the catalytic nucleophile, and glutamic acid 188 is a candidate for the acid/base catalyst. These residues are located in the fourth and seventh β-strand of the TIM barrel domain, respectively. Three isoforms of β-galactosidase have been described that are produced by alternative splicing.  Isoform 1 is present in lysosomes and isoform 2 is detected in the cytoplasm and in perinuclear region. Isoform 2 lacks β-galactosidase catalytic activity but it plays a functional role in the formation of extracellular elastic fibers and in the development of connective tissue.  Defects in and dysfunction of β-galactosidase  and mutations in GLB1 gene have been linked to metabolic storage disorders, such a GM1 gangliosidosis, a severe neurological disease. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Ohtu, U., et al. (2012). J. Biol. Chem. 287(3); 1801-1812; Caciotti, A., et al. (2005). Hum. Mutat. 25(3); 285-292).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

0.3 mg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service