Skip to Content
Merck
All Photos(13)

Documents

ZRB1583

Sigma-Aldrich

Anti-PDE4B Antibody, clone 1E9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

5′-cyclic phosphodiesterase 4B, DPDE4, EC:3.1.4.53, PDE32, cAMP-specific 3′

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E9, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 83.34 kDa
observed mol wt ~100 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PDE4B(5142)
mouse ... Pde4B(18578)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1E9 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects cAMP-specific 3′,5′-cyclic phosphodiesterase 4B (PDE4B). It targets an epitope within 22 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal region of human cAMP-specific 3′,5′-cyclic phosphodiesterase 4B (PDE4B).

Application

Anti-PDE4B, clone 1E9 ZooMAb, Cat. No. ZRB1583, is a recombinant Rabbit monoclonal antibody that specifically targets PDE4B and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PDE4B in lysates from HL-60, HT-29, and HepG2 cells and mouse brain tissue lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PDE4B in HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PDE4B in human placenta tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound PDE4B peptide with a KD of 1.2 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

cAMP-specific 3′,5′-cyclic phosphodiesterase 4B (UniProt: Q07343; also known as EC:3.1.4.53, DPDE4, PDE32) is encoded by the PDE4B (also known as DPDE4) gene (Gene ID: 5142) in human. Cyclic nucleotide phosphodiesterases (PDEs) play a critical role in the regulation of cAMP signaling. They degrade and inactivate cAMP and terminate its action. PDE4B is expressed in brain, heart, lung and skeletal muscle. It is also the predominant PDE subtype in monocytes and neutrophils. Two variants of PDE4B (PDE4B1 and PDE4B2) have been described in human and rat. Its cAMP binding site of PDE4B is localized in amino acids 406-410. PDE4B is shown to be crucial in the regulation of cardiac excitation-contraction coupling (ECC) and its inhibition increases the tendency of cardiac arrhythmias. It is reported to function in a confined compartment near the sarcolemma of cardiac myocytes and it controls subsarcolemmal cAMP but not the cytosolic cAMP. A reduction in PDE4B expression and activity has been observed during cardiac hypertrophy. Four isoforms of PDE4B have been described that are produced by alternative splicing. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Mika, D., et al. (2014). J. Cell Sci. 127(5); 1033-1042).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service