Skip to Content
Merck
All Photos(2)

Key Documents

WH0389692M1

Sigma-Aldrich

Monoclonal Anti-MAFA antibody produced in mouse

clone 2F1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-RIPE3b1, Anti-hMafA, Anti-v-maf musculoaponeurotic fibrosarcoma oncogene homolog A (avian)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAFA(389692)

General description

MAFA is a transcription factor that binds RIPE3b, a conserved enhancer element that regulates pancreatic beta cell-specific expression of the insulin gene (INS; MIM 176730) (Olbrot et al., 2002 [PubMed 12011435]).[supplied by OMIM

Immunogen

MAFA (NP_963883, 222 a.a. ~ 308 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LEERFSDDQLVSMSVRELNRQLRGFSKEEVIRLKQKRRTLKNRGYAQSCRFKRVQQRHILESEKCQLQSQVEQLKLEVGRLAKERDL

Biochem/physiol Actions

MAF bZIP transcription factor A (MAFA) modulates genes which are involved in pancreatic β-cell function and the protein itself is critical for the functioning of β-cells. It functions as a regulator to enhance the expression of genes which have a role in glucose-induced insulin secretion.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan L Vanderford
Islets, 3(1), 35-37 (2011-02-01)
MafA, a basic-leucine zipper transcription factor that is important to pancreatic β-cell function, is regulated by several intricate mechanisms. MafA undergoes extensive posttranslational modification by phosphorylation, ubiquitination and sumoylation, and these modifications regulate the turnover, DNA binding and transactivation function
A E Butler et al.
Diabetologia, 55(11), 2985-2988 (2012-08-01)
The beta cell transcriptional factor musculoaponeurotic fibrosarcoma oncogene family A (MafA) regulates genes important for beta cell function. Loss of nuclear MafA has been implicated in beta cell dysfunction in animal models of type 2 diabetes. We sought to establish

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service