Skip to Content
Merck
All Photos(1)

Documents

SRP0465

Sigma-Aldrich

BRDT (257-382) His tag human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

BRD6, CT9, bromodomain, testis-specific

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

15.8 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRDT(676)

General description

Human Bromodomain testis-specific protein, also known as BRDT, BRD6, and CT9 (GenBank Accession No. NM_207189) amino-acids 257 – 382 with N-terminal HIS-tag, MW = 15.8 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binyamin D Berkovits et al.
Developmental biology, 360(2), 358-368 (2011-10-25)
Mice homozygous for a mutation (Brdt(∆BD1/∆BD1)) lacking the first bromodomain of Brdt, a testis-specific member of the BET family of double-bromodomain containing proteins, are sterile and exhibit profound defects in chromatin remodeling during spermiogenesis. We have now observed that a
Enyuan Shang et al.
Development (Cambridge, England), 134(19), 3507-3515 (2007-08-31)
Brdt is a testis-specific member of the distinctive BET sub-family of bromodomain motif-containing proteins, a motif that binds acetylated lysines and is implicated in chromatin remodeling. Its expression is restricted to the germ line, specifically to pachytene and diplotene spermatocytes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service