Skip to Content
Merck
All Photos(2)

Key Documents

SRP0434

Sigma-Aldrich

PDE7A active mouse

recombinant, expressed in baculovirus infected Sf9 cells, ≥22% (SDS-PAGE)

Synonym(s):

phosphodiesterase 7A

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$687.00

$687.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$687.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$687.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

mouse

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥22% (SDS-PAGE)

form

aqueous solution

mol wt

65 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... PDE7A(18583)

General description

Mouse PDE7A (GenBank Accession No. NM_001122759) amino acids 147-482 (end) with N-terminal GST-tag, MW=65 kDa, expressed in a Baculovirus-infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

1 unit is defined as the amount of enzyme that will convert 1 pmole of 3?, 5?-cAMP to 5? AMP per min at 37°C.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Yamamoto et al.
European journal of pharmacology, 541(1-2), 106-114 (2006-06-20)
YM-393059, (+/-)-N-(4,6-dimethylpyrimidin-2-yl)-4-[2-(4-methoxy-3-methylphenyl)-5-(4-methylpiperazin-1-yl)-4,5,6,7-tetrahydro-1H-indol-1-yl]benzenesulfonamide difumarate, is a novel phosphodiesterase (PDE) inhibitor that inhibited the PDE7A isoenzyme with a high potency (IC50=14 nM) and PDE4 with a moderate potency (IC50=630 nM). In a cell-based assay, YM-393059 was found to inhibit anti-CD3 antibody, Staphylococcal
Guchen Yang et al.
Journal of immunology (Baltimore, Md. : 1950), 171(12), 6414-6420 (2003-12-10)
Phosphodiesterases (PDEs) are enzymes which hydrolyze the cyclic nucleotide second messengers, cAMP and cGMP. In leukocytes, PDEs are responsible for depletion of cAMP which broadly suppresses cell functions and cellular responses to many activation stimuli. PDE7A has been proposed to
Assignment of the mouse Pde7A gene to the proximal region of chromosome 3 and of the human PDE7A gene to chromosome 8q13.
P Han et al.
Genomics, 48(2), 275-276 (1998-04-02)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service