Skip to Content
Merck
All Photos(2)

Key Documents

SML3883

Sigma-Aldrich

Cephalofurimazine

≥98% (HPLC)

Synonym(s):

8-benzyl-6-(2,3-difluorophenyl)-2-(furan-2-ylmethyl)imidazo[1,2-a]pyrazin-3(7H)-one, CFz

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H17F2N3O2
Molecular Weight:
417.41
UNSPSC Code:
41106609
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Brain penetrant NanoLuc luciferase substrate with improved brain performance.

Cephalofurimazine is a brain penetrant NanoLuc luciferase substrate with improved brain performance. Cephalofurimazine combined with Antares luciferase generates greater than 20-fold more signal from the brain than standard d-luciferin with firefly luciferase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An optimized bioluminescent substrate for non-invasive imaging in the brain
Nature Chemical Biology, 19(6), 731-739 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service