Skip to Content
Merck
All Photos(1)

Key Documents

SML3244

Sigma-Aldrich

CU-CPD107

≥98% (HPLC)

Synonym(s):

2-(Ethoxymethyl)-4-iodo-α,α-dimethyl-5-phenyl-1H-imidazole-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21IN2O2
CAS Number:
Molecular Weight:
400.25
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C)(O)CN(C(COCC)=N1)C(C2=CC=CC=C2)=C1I

InChI

1S/C16H21IN2O2/c1-4-21-10-13-18-15(17)14(12-8-6-5-7-9-12)19(13)11-16(2,3)20/h5-9,20H,4,10-11H2,1-3H3

Biochem/physiol Actions

CU-CPD107 is a selective and potent inhibitor of Resiquimod-induced Toll-like receptor 8 (TLR8) signaling. Additionally, CU-CPD107 acts as TLR8 agonist in the presence of ssRNA. CU-CPD107 alone is unable to modulate TLR8 signaling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Yang et al.
Nature communications, 12(1), 4351-4351 (2021-07-18)
Small-molecule modulators of TLR8 have drawn much interests as it plays pivotal roles in the innate immune response to single-stranded RNAs (ssRNAs) derived from viruses. However, their clinical uses are limited because they can invoke an uncontrolled, global inflammatory response.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service