Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702567

Sigma-Aldrich

Anti-SOX1 Antibody, clone 9L8Z4, Rabbit Monoclonal

Synonym(s):

SRY (sex determining region Y)-box 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

9L8Z4, monoclonal

form

liquid

mol wt

45 kDa

species reactivity

rat, rat, human

concentration

1 mg/mL

technique(s)

western blot: 1:1000 - 1:2200

color

colorless

isotype

IgG

immunogen sequence

MYSMMMETDLHSPGGAQAPTNLSGPAGAGGGGGGGGGGGGGGGAKANQDRVKRPMNAFMVWSRGQRRKMAQENPKMHNSEISKRLGAEWKVMSEAEKRPF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX1(6656)

General description

This intronless gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. In mice, a similar protein regulates the gamma-crystallin genes and is essential for lens development. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SOX1 (O00570).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service