Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200865

Sigma-Aldrich

Anti-Pro GDF11 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

GDF11 Growth/differentiation factor 11 Bonemorphogenetic protein11 (bmp11)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

antibody form

affinity isolated antibody

Quality Level

form

liquid

species reactivity

mouse, rat

concentration

~1 mg/mL

technique(s)

immunoblotting: 1:500-1:1000 using rat PC12 cell line

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Growth Differentiation Factor 11 (GDF11) is expressed in many tissues, including skeletal muscle, pancreas, kidney, nervous system, and retina, among others. GDF11 circulating levels and protein content in tissues are quite variable and are affected by pathological conditions or age.1 GDF11 also known as Bone Morphogenetic Protein 11 (BMP-11), mRNA is translated in a precursor protein, which is processed by specific proteases generating the mature GDF11 (C-terminal, 12.5 kDa) and the pro-domain (Nterminal, 30.1 kDa). GDF11 shares 89% amino acid sequence homology with GDF8, however GDF8 expression in human tissues is restricted to cardiac and skeletal muscle,2 while GDF11 is expressed in all tissues.3 Although there is high homology between mature GDF8 and GDF11, the pro-domains of both proteins share only 54% homology. The pro-domain is fundamental for proper protein folding, disulfide bond formation and exportation of the homodimers,4,5 suggesting differences in posttranslational process.

Specificity

Anti-Pro GDF11 antibody specifically recognizes Pro GDF11 from human origin.

Application

The antibody may be used in various immunochemical techniques including Immunoblotting. Detection of the Pro GDF11 band by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

GDF11 uses the canonical receptors and the SMAD proteins for signaling. The GDF11 dimer (a disulfide-linked homodimer of carboxy-terminal fragments) binds the activin receptors type II A or B (ActRIIA, ActRIIB), proteins with serine/threonine kinase activity.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco S Loffredo et al.
Cell, 153(4), 828-839 (2013-05-15)
The most common form of heart failure occurs with normal systolic function and often involves cardiac hypertrophy in the elderly. To clarify the biological mechanisms that drive cardiac hypertrophy in aging, we tested the influence of circulating factors using heterochronic
Mathias Uhlén et al.
Science (New York, N.Y.), 347(6220), 1260419-1260419 (2015-01-24)
Resolving the molecular details of proteome variation in the different tissues and organs of the human body will greatly increase our knowledge of human biology and disease. Here, we present a map of the human tissue proteome based on an
A Jamaiyar et al.
Pharmacology & therapeutics, 175, 28-34 (2017-02-23)
In addition to its roles in embryonic development, Growth and Differentiation Factor 11 (GDF 11) has recently drawn much interest about its roles in other processes, such as aging. GDF 11 has been shown to play pivotal roles in the
A M Gray et al.
Science (New York, N.Y.), 247(4948), 1328-1330 (1990-03-16)
Many proteins are initially synthesized as part of a large precursor. The role of the pro-region in the biosynthesis of transforming growth factor--beta 1 (TGF-beta 1) and activin A, two structurally related disulfide-linked homodimers synthesized as large precursors, was studied.
Arturo Simoni-Nieves et al.
Frontiers in oncology, 9, 1039-1039 (2019-11-05)
Growth Differentiation Factor 11 (GDF11), a member of the super family of the Transforming Growth Factor β, has gained more attention in the last few years due to numerous reports regarding its functions in other systems, which are different to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service