Skip to Content
Merck
All Photos(1)

Documents

SAB4200189

Sigma-Aldrich

Anti-Sumo-1 antibody, Rat monoclonal

clone 4D12, purified from hybridoma cell culture

Synonym(s):

Anti-DAP1, Anti-GMP1, Anti-OFC10 CLEFT LIP WITH OR WITHOUT CLEFT PALATE, NONSYNDROMIC, 10, Anti-SMT3 suppressor of mif two 3 homolog 1 SENTRIN PIC1 OROFACIAL CLEFT 10, INCLUDED; OFC10, Anti-SMT3H3 SMT3C, Anti-UBL1, SMT3, YEAST, HOMOLOG 3, Anti-small ubiquitin-like modifier 1, Anti-ubiquitin-like 1, Monoclonal Anti-Sumo-1, SUMO-1, SUMO1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

4D12, monoclonal

form

buffered aqueous solution

mol wt

~12 kDa

species reactivity

human, rat, mouse

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: 2-4 μg/mL using HeLa cell extracts

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SUMO1(7341)
mouse ... Sumo1(22216)
rat ... Sumo1(301442)

General description

Monoclonal Anti-Sumo-1 (rat IgG2a isotype) is derived from the hybridoma 4D12 produced by the fusion of mouse myeloma cells (SP2/0) and splenocytes from rat immunized with a fusion protein of human SUMO1.
SUMO1 (small ubiquitin-related modifier), also called as Pic1 (inner centromere protein-related protein), Ubl1 (ubiquitin-like protein), hSmt3 (human homolog 3), or sentrin, is mainly expressed in the nucleus. The gene encoding SUMO1 is mapped to human chromosome 2q33. SUMO1 belongs to ubiquitin-like protein family and it consists of 101 amino acids.

Immunogen

fusion protein of human SUMO1

Application

Monoclonal Anti-Sumo-1 has been used in immunocytochemistry.
Monoclonal Anti-Sumo-1 has been used in western blotting.

Biochem/physiol Actions

Sumo-1 interacts with dsDNA and competes with other protein-DNA interactions.
Suppressing SUMO1 (small ubiquitin-related modifier) inhibits multiplication and induces apoptosis of gastric cancer SGC-7901 cells. It is essential for HCV (hepatitis C virus) replication. Deletion of SUMO1 blocks the aggregation of lipid droplets. It is also implicated in mitochondrial fission.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SUMO-1 possesses DNA binding activity
Eilebrecht S, et al.
BMC Research Notes, 3(1), 146-146 (2010)
Donatella Ponti et al.
PloS one, 9(5), e96037-e96037 (2014-05-03)
EGR1 is an immediate early gene with a wide range of activities as transcription factor, spanning from regulation of cell growth to differentiation. Numerous studies show that EGR1 either promotes the proliferation of stimulated cells or suppresses the tumorigenic growth
The genetic basis of cleft lip and cleft palate
Veros C
Aristotle University Medical Journal, 43, 25-36 (2016)
SUMO-1 Gene Silencing Inhibits Proliferation and Promotes Apoptosis of Human Gastric Cancer SGC-7901 Cells.
Jin L
Cellular Physiology and Biochemistry, 41, 987-998 (2017)
Sumo1 conjugates mitochondrial substrates and participates in mitochondrial fission.
Harder Z
Current Biology, 14, 340-345 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service