Skip to Content
Merck
All Photos(2)

Documents

SAB2102059

Sigma-Aldrich

Anti-RTCD1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-RNA terminal phosphate cyclase domain 1, Anti-RPC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

dog, horse, guinea pig, rabbit, human, mouse, rat, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RTCD1(8634)

Immunogen

Synthetic peptide directed towards the N terminal region of human RTCD1

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

RNA 3-prime-terminal phosphate cyclase catalyzes the ATP-dependent conversion of a 3-prime phosphate to a 2-prime,3-prime-cyclic phosphodiester at the end of RNA.RNA 3-prime-terminal phosphate cyclase (RPC; EC 6.5.1.4) catalyzes the ATP-dependent conversion of a 3-prime phosphate to a 2-prime,3-prime-cyclic phosphodiester at the end of RNA (Genschik et al., 1997 [PubMed 9184239]).[supplied by OMIM]. Sequence Note: removed 1 base from the 3′ end that did not align to the reference genome assembly. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-1538 Y11651.1 1-1538

Sequence

Synthetic peptide located within the following region: VQKIRAGRSTPGLRPQHLSGLEMIRDLCDGQLEGAEIGSTEITFTPEKIK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanquan Song et al.
Nature neuroscience, 18(6), 817-825 (2015-05-12)
Mechanisms governing a neuron's regenerative ability are important but not well understood. We identify Rtca (RNA 3'-terminal phosphate cyclase) as an inhibitor of axon regeneration. Removal of Rtca cell-autonomously enhanced axon regrowth in the Drosophila CNS, whereas its overexpression reduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service