Skip to Content
Merck
All Photos(4)

Key Documents

SAB1402084

Sigma-Aldrich

Monoclonal Anti-SLC13A5 antibody produced in mouse

clone 2G4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp686E17257, MGC138356, NACT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2G4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The solute carrier family 13 member 5 (SLC13A5) gene is mapped to human chromosome 17p13.1. It is highly expressed in the plasma membrane of hepatocytes and is also expressed in the cells of testis and brain.

Immunogen

SLC13A5 (NP_808218, 152 a.a. ~ 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VEAILQQMEATSAATEAGLELVDKGKAKELPGSQVIFEGPTLGQQEDQERKRLCK

Biochem/physiol Actions

SLC13A5 (solute carrier family 13 member 5) is a sodium-coupled citrate transporter. Its function involves the transportation of citrate from the bloodstream into the liver. The expression of the gene is found to be increased in obesity and non-alcoholic fatty liver disease. Downregulation of SLC13A5 promotes AMPK (adenosine monophosphate-activated protein kinase) activation and inhibits ACLY (ATP citrate lyase) expression. SLC13A5 is thus found to regulate homeostasis of energy and lipid in liver cancer, as the gene is associated with synthesis of fatty acids and cholesterol. Mutation in the SLC13A5 gene leads to early occurrence of epilepsy, developmental delay and tooth dysplasia in children.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flipping a citrate switch on liver cancer cells.
Peters JM
The Journal of Biological Chemistry, 292(33), 13902-13903 (2017)
Silencing of solute carrier family 13 member 5 disrupts energy homeostasis and inhibits proliferation of human hepatocarcinoma cells.
Li Z
The Journal of Biological Chemistry, 292(33), 13890-13901 (2017)
Mutations in the Na(+)/citrate cotransporter NaCT (SLC13A5) in pediatric patients with epilepsy and developmental delay.
Klotz J
Molecular Medicine, 22, 1-12 (2016)
Epilepsy and mental retardation restricted to females: X-linked epileptic infantile encephalopathy of unusual inheritance.
Duszyc K
Journal of applied genetics, 56(1), 49-56 (2015)
Plasma Membrane xn-Na-zbu-Coupled Citrate Transporter (SLC13A5) and Neonatal Epileptic Encephalopathy
Bhutia YD, et al.
Molecules (Basel), 3-3 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service