Skip to Content
Merck
All Photos(1)

Documents

S0501

Sigma-Aldrich

Sodium nitroprusside dihydrate

Synonym(s):

Nitroprusside sodium, SNP, Sodium nitroferricyanide, Sodium pentacyanonitrosylferrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
CAS Number:
Molecular Weight:
297.95
EC Number:
MDL number:
UNSPSC Code:
12352300

SMILES string

O.O.[Na+].[Na+].O=N[Fe--](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/5CN.Fe.NO.2Na.2H2O/c5*1-2;;1-2;;;;/h;;;;;;;;;2*1H2/q;;;;;2*-1;2*+1;;

InChI key

OIRZWVYIQXBRFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Releases NO in vivo, thus activating guanylyl cyclase, ADP-ribosyltransferase, and cyclooxygenase and inhibiting lipoxygenase. Induces vasodilation and inhibits platelet aggregation.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Brüne et al.
The Journal of biological chemistry, 264(15), 8455-8458 (1989-05-25)
Sodium nitroprusside is a vasodilator and an inhibitor of platelet activation. It is thought that these effects are mediated by the spontaneous release of nitric oxide and stimulation of cytosolic guanylate cyclase. We have found that sodium nitroprusside (5-200 microM)
Feelisch, M.
Journal of Cardiovascular Pharmacology, 17, S25-S25 (1991)
M Maccarrone et al.
Biochemical and biophysical research communications, 219(1), 128-133 (1996-02-06)
The nitric oxide (N0-releasing agents sodium nitroprusside (SNP) and S-nitroso-N-acetylpenicillamine (SNAP) inhibit dioxygenase activity of lipoxygenase in human platelets and human CHP100 neuroblastoma cells, leading the latter to necrosis. The effect of both NO-donors on the dioxygenase reaction was investigated
D Salvemini et al.
The Journal of clinical investigation, 97(11), 2562-2568 (1996-06-01)
We have evaluated the contributions of nitric oxide (NO) and prostacyclin (PGI2) in the in vivo antiplatelet effects of clinically useful nitrovasodilators. In rats, intravenous infusion of three NO donors, glyceryl trinitrate, sodium nitroprusside, or 3'-morpholinosydnonimine, the stable metabolite of
C M Roos et al.
Journal of applied physiology (Bethesda, Md. : 1985), 77(1), 51-57 (1994-07-01)
We localized the sites of vasodilation of inhaled nitric oxide (NO), a selective pulmonary vasodilator, and sodium nitroprusside (SNP) in isolated rat lungs. The sites were determined by analyzing the arterial, venous, and double-occlusion data with a two-resistor (small arteries

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service