OH30429807
Ohaus® Pioneer® Precision Balance
Model PX3202, weighing capacity 3200 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
metal weighing plate
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30429807
weighing capacity measuring range
3200 g, resolution: 0.01 g
weighing pan diam.
18 cm
Looking for similar products? Visit Product Comparison Guide
General description
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. The PX is affordably priced, and intuitively designed for intelligent operation with a second line display for additional information, and USB and RS232 connectivity for easy communication.
Application
Basic Weighing, Parts Counting, Percent Weighing, Animal Weighing, Density Determination
Features and Benefits
User-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, userselectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator, four operating languages
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Pioneer is a registered trademark of Ohaus Corp.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service