Skip to Content
Merck
All Photos(1)

Key Documents

O7628

Sigma-Aldrich

Eupergit® C

~150 μm (macroporous particles)

Synonym(s):

Copolymer of methacrylamide, N,N′-methylen-bis(acrylamide) and a monomer carrying oxirane groups, Epoxide polymer-bound, Oxirane acrylic beads

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
13111000

extent of labeling

~800 μmol per g

matrix spacer

3 atoms (when ligands are coupled through the free oxirane groups. Linkage is electroneutral.)

particle size

~150 μm (macroporous particles)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Matrix: hydrophilic acrylic beads

Legal Information

Eupergit is a registered trademark of Röhm GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michiel H A Janssen et al.
Biotechnology and bioengineering, 78(4), 425-432 (2002-04-12)
Penicillin G acylase from Escherichia coli was immobilized on Eupergit C with different enzyme loading. The activity of the immobilized preparations was assayed in the hydrolysis of penicillin G and was found to be much lower than would be expected
Salwa Karboune et al.
Journal of biotechnology, 120(3), 273-283 (2005-08-10)
Chlorophyllase extract from Phaeodactylum tricornutum was immobilized by physical adsorption on DEAE-cellulose and silica gel as well as by covalent binding on Eupergit C, Eupergit C250L, Eupergit C/ethylenediamine (EDA) and Eupergit C250L/EDA. Although the highest immobilization yield (83-93%) and efficiency
S S Tan et al.
Bioresource technology, 99(1), 200-204 (2007-01-30)
In this study, a thermostable recombinant xylanase B (XynB) from Thermotoga maritima MSB8 was immobilized on nickel-chelated Eupergit C 250L. This immobilized XynB was then used to hydrolyze the autohydrolysis explosion liquor of corncob (AELC) in a packed-bed enzyme reactor
Caterina Temporini et al.
Biomacromolecules, 11(6), 1623-1632 (2010-05-14)
An innovative approach to determine the orientation of penicillin G acylase (PGA) from Escherichia coli covalently immobilized onto solid supports has been developed. This method is based on tryptic digestion of immobilized PGA followed by HPLC-MS analysis of the released
Rihui Lin et al.
Preparative biochemistry & biotechnology, 41(2), 154-165 (2011-03-29)
Oxalate decarboxylase, an oxalate degradation enzyme used for medical diagnosis and decreasing the oxalate level in the food or paper industry, was covalently immobilized to Eupergit C. Different immobilization parameters, including ratio of enzyme to support, ammonia sulfate concentration, pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service