Skip to Content
Merck
All Photos(1)

Documents

N3135

Supelco

Nalorphine hydrochloride

Synonym(s):

N-Allylnormorphine HCl, Nalline HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO3 · HCl
CAS Number:
Molecular Weight:
347.84
EC Number:
MDL number:
UNSPSC Code:
41116107

drug control

USDEA Schedule III; regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

H2O: slightly soluble
dilute aqueous acid: soluble
ethanol: soluble

SMILES string

Cl[H].O[C@H]1C=CC2[C@H]3Cc4ccc(O)c5O[C@@H]1[C@]2(CCN3CC=C)c45

InChI

1S/C19H21NO3.ClH/c1-2-8-20-9-7-19-12-4-6-15(22)18(19)23-17-14(21)5-3-11(16(17)19)10-13(12)20;/h2-6,12-13,15,18,21-22H,1,7-10H2;1H/t12?,13-,15+,18+,19+;/m1./s1

InChI key

NAHATSPWSULUAA-HWXFZQNOSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Antagonist at μ receptors, partial agonist at κ receptors, and agonist at sigma receptors, but does not have high affinity for the sigma receptor

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C D King et al.
Archives of biochemistry and biophysics, 332(1), 92-100 (1996-08-01)
Rat and human UDP-glucuronosyltransferase (UGT) 1.1 share > 70% identity in their deduced primary amino acid sequences. We have previously shown that rat UGT1.1, stably expressed in human embryonic kidney 293 cells, catalyzes the glucuronidation of bilirubin and the mixed
David A White et al.
The Journal of pharmacology and experimental therapeutics, 314(1), 374-382 (2005-04-22)
The discriminative stimulus effects of acute morphine followed by naltrexone have been described previously in nonhuman primates. The purposes of this study were to 1) extend the pharmacological characterization of the discrimination by testing mu-opioid agonists other than morphine and
C D King et al.
Drug metabolism and disposition: the biological fate of chemicals, 25(2), 251-255 (1997-02-01)
Opioids are important drugs used as analgesics, antitussives, antidiarrheals, and in the therapy of myocardial infarctions, and as antagonists of opioid intoxication. The glucuronidation of these compounds, catalyzed by UDP-glucuronosyltransferases (UGTs), is well known to be a primary step in
Mark A Smith et al.
Pharmacology, biochemistry, and behavior, 76(1), 93-101 (2003-09-19)
Previous studies have reported that social and environmental enrichment can have a marked impact on the functional maturation of the central nervous system and may influence an organism's sensitivity to psychotropic drugs. The purpose of the present study was to
Andrew C Barrett et al.
Psychopharmacology, 160(2), 170-181 (2002-03-05)
Recent studies indicate that sex and rodent strain are determinants of sensitivity to opioid-induced antinociception. The present study examined the influence of sex and rat strain on kappa opioid-induced antinociception using a series of kappa opioids that vary in their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service