Skip to Content
Merck
All Photos(1)

Key Documents

M8569

Sigma-Aldrich

Anti-MAFF (N-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-v-maf Avian Musculoaponeurotic Fibrosarcoma Oncogene Family, ProteinF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 18 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): 4-8 μL using A431 cell line
western blot: 1:4,000-1:8,000 using whole extracts of the A431 cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAFF(23764)
mouse ... Maff(17133)
rat ... Maff(366960)

General description

Anti-MAFF (N-terminal) is developed in rabbit using a synthetic peptide corresponding to amino acids of human MAFF, conjugated to keyhole limpet hemocyanin (KLH) via a C-terminal added cysteine residue, as immunogen. The Maf family is divided into two subclasses, large Mafs (vMaf, cMaf, MafB and Nrl) and small Mafs (MafF, MafK, and MafG). Human MAF bZIP transcription factor F (MAFF) encodes a 164 amino acids protein. It lacks an N-terminal transactivating domain.
v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F (MAFF) is a transcription factor which does not have a transactivation domain but possesses a leucine-zipper.

Immunogen

synthetic peptide corresponding to amino acids 2-17 of human MAFF, conjugated to KLH via a C-terminal added cysteine residue. The mouse and rat sequences differ from the human by one and two amino acids, respectively. Seven amino acids are identical in MaFG in the C-terminal region of the peptide.

Application

Anti-MAFF (N-terminal) antibody produced in rabbit has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

The three small Maf proteins participates in development, differentiation, hematopoiesis and stress response.
v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F (MAFF) has been shown to interact with other transcription factors and it associates with certain regulatory sequences in the gene encoding the oxytocin receptor.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Minimal Transactivation Domain of the Basic Motif-Leucine Zipper Transcription Factor NRL Interacts with TATA-binding Protein
Friedman JS, et al.
The Journal of Biological Chemistry, 279(45), 47233-47241 (2004)
Differential induction of mafF, mafG and mafK expression by electrophile-response-element activators
Moran JA, et al.
The Biochemical Journal, 361(2), 371-377 (2002)
O Johnsen et al.
Nucleic acids research, 24(21), 4289-4297 (1996-11-01)
The human TCF11 gene encodes a ubiquitously expressed bZIP transcription factor of the cap n' collar (CNC) domain family. It has a high sequence similarity to the erythroid-specific bZIP factor p45 NF-E2 in the CNC domain, which is involved in
Wael Massrieh et al.
Biology of reproduction, 74(4), 699-705 (2005-12-24)
The MAF (proto-)oncogene family of basic-leucine zipper transcription factors plays crucial roles in the control of mammalian gene expression and development. Here we analyzed the regulation of the human MAFF gene, coding for a small MAF transcription factor, in uterine
Xiaoxia Ye et al.
Archives of biochemistry and biophysics, 449(1-2), 87-93 (2006-03-22)
The human transcription factor MafF (hMafF) lacks a transactivation domain, it contains a heptad-leucine repeat motif (viz., Leu-zipper) that may mediate protein-protein interactions to regulate transcriptional activities. A protein with a coiled-coil domain encoded by a novel human gene (GenBank

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service