Skip to Content
Merck
All Photos(1)

Documents

M0135

Sigma-Aldrich

Myristoyl-DL-carnitine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H42NO4Cl
CAS Number:
Molecular Weight:
408.02
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cl.CCCCCCCCCCCCCC(=O)OC(CC(O)=O)C[N](C)(C)C

InChI

1S/C21H42NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4;/h19H,5-18H2,1-4H3,(H,23,24);1H

InChI key

CDEZWHYFAXNHSK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lawrence C Johnson et al.
Clinical science (London, England : 1979), 132(16), 1765-1777 (2018-06-20)
Advancing age is associated with impairments in numerous physiological systems, leading to an increased risk of chronic disease and disability, and reduced healthspan (the period of high functioning healthy life). The plasma metabolome is thought to reflect changes in the
Arif Yurdagul et al.
Cell metabolism, 31(3), 518-533 (2020-02-01)
Continual efferocytic clearance of apoptotic cells (ACs) by macrophages prevents necrosis and promotes injury resolution. How continual efferocytosis is promoted is not clear. Here, we show that the process is optimized by linking the metabolism of engulfed cargo from initial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service