Skip to Content
Merck
All Photos(1)

Documents

I8255

Sigma-Aldrich

19-Iodocholesterol 3-acetate

~95% (TLC)

Synonym(s):

19-Iodo-5-cholesten-3β-ol 3-acetate, 5-Cholesten-19-iodo-3β-ol 3-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H47IO2
CAS Number:
Molecular Weight:
554.59
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

~95% (TLC)

storage temp.

−20°C

SMILES string

IC[C@@](C1=CC2)(CC[C@H](OC(C)=O)C1)[C@]3([H])[C@]2([H])[C@@](CC[C@]4([H])[C@H](C)CCCC(C)C)([H])[C@]4(C)CC3

InChI

1S/C29H47IO2/c1-19(2)7-6-8-20(3)25-11-12-26-24-10-9-22-17-23(32-21(4)31)13-16-29(22,18-30)27(24)14-15-28(25,26)5/h9,19-20,23-27H,6-8,10-18H2,1-5H3/t20-,23+,24+,25-,26+,27+,28-,29-/m1/s1

InChI key

LVQSCLBKGHGMFF-KVHJHAPGSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Owoyale
Steroids, 38(2), 205-209 (1981-08-01)
Since 19-iodocholesterol ethyl and methyl ethers and 3 beta-iodo-5-cholestene localized in dog adrenal, the distribution of 19-iodocholesterol 3-acetate was similarly studied. This ester was concentrated by the dog adrenal to the same extent as 19-iodocholesterol. Despite a previous report that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service