Skip to Content
Merck
All Photos(1)

Documents

HPA044681

Sigma-Aldrich

Anti-FFAR3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FFA3R, Anti-GPR41, Anti-free fatty acid receptor 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

DFHELLRRLCGLWGQWQQESSMELKEQKGGEEQRADRPAERKTSEHSQGCGTGGQVAC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FFAR3(2865)

General description

FFAR3 (free fatty acid receptor 3) is also known as G-protein-coupled receptor 41 (GPR41). It is a 346 amino acid receptor that has similar structure like GPR43.It is present on enteroendocrine L cells. This gene is mapped to human chromosome 19q13.1.

Immunogen

free fatty acid receptor 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-FFAR3 antibody has been used in immunohistochemistry.

Biochem/physiol Actions

FFAR3 may participate in the acetylation-related modulation of cell processes like proliferation, apoptosis and the cell cycle. Short-chain fatty acids (C2–C5) induces GPR41, that helps to link to Gai, which blocks the adenylyl cyclase pathway attenuating cAMP synthesis. It performs various functions in vivo, such as immune response and energy homoeostasis. GPR41 promotes the liberation of GLP-1 and PYY and on pancreatic beta cells, it may block the liberation of insulin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83874

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Butyrate-induced GPR41 activation inhibits histone acetylation and cell growth.
Wu J, et al.
Journal of Genetics and Genomics = Yi Chuan Xue Bao, 39(8), 375-384 (2012)
G protein-coupled receptors: signalling and regulation by lipid agonists for improved glucose homoeostasis.
Moran B M, et al.
Acta Diabetologica, 53(2), 177-188 (2016)
Zhiwei Ang et al.
Scientific reports, 6, 34145-34145 (2016-09-27)
Knockout mice studies implicate the mammalian short-chain fatty acid (SCFA) receptors, FFAR2 and FFAR3- in colitis, arthritis and asthma. However, the correlation with human biology is uncertain. Here, we detected FFAR2 and FFAR3 expression in human monocytes via immunohistochemistry. Upon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service