Skip to Content
Merck
All Photos(2)

Documents

HPA038941

Sigma-Aldrich

Anti-NEK10 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ32685, Anti-NIMA (never in mitosis gene a)- related kinase 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LHILQGDRNFVSDHSSIGSLSSANAAGRIQQLHLSEDLSPREIQENTFSLQAACCAALTELVLNDTNAHQVVQENGVYTIAKLILPNKQKNAAKSNLLQCYAFRALRFLFSMERNRPLFKRLFPTDLF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEK10(152110)

General description

The gene NEK10 (NIMA related kinase 10) is mapped to human chromosome 3p24. The encoded protein belongs to the NEK kinase family of proteins. NEK10 is a serine threonine kinase. The protein has armadillo repeats, coiled coil domains, a kinase domain and a PEST (proline (P), glutamic acid (E), serine (S), and threonine (T)) sequence. NEK10 mRNA is present in the lung, spleen and mammary glands.

Immunogen

NIMA (never in mitosis gene a)- related kinase 10 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NEK10 (NIMA related kinase 10) regulates G2/M checkpoint after ultraviolet irradiation. It positively regulates ERK1/2 (extracellular signal-regulated kinase) signaling. Genome-wide association study has shown association between NEK10 and breast cancer. Mutations in NEK10 gene have been reported in lung cancer cell lines.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71470

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irene García-Díez et al.
Genes, chromosomes & cancer, 58(3), 164-174 (2018-11-27)
Although most cutaneous squamous cell carcinomas (cSCCs) develop from actinic keratoses (AKs), the key events in this evolution remain unclear. We have combined the results of different genomic and expression array platforms on matched concomitant samples of sun-exposed skin (SES)
Nek10 mediates G2/M cell cycle arrest and MEK autoactivation in response to UV irradiation.
Moniz LS and Stambolic V
Molecular and Cellular Biology, 31, 30-42 (2011)
Irene García-Díez et al.
Genes, chromosomes & cancer, 58(3), 164-174 (2018-11-27)
Although most cutaneous squamous cell carcinomas (cSCCs) develop from actinic keratoses (AKs), the key events in this evolution remain unclear. We have combined the results of different genomic and expression array platforms on matched concomitant samples of sun-exposed skin (SES)
Raghu R Chivukula et al.
Nature medicine, 26(2), 244-251 (2020-01-22)
Mucociliary clearance, the physiological process by which mammalian conducting airways expel pathogens and unwanted surface materials from the respiratory tract, depends on the coordinated function of multiple specialized cell types, including basal stem cells, mucus-secreting goblet cells, motile ciliated cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service