Skip to Content
Merck
All Photos(1)

Documents

HPA025062

Sigma-Aldrich

Anti-SLCO5A1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-OATP-RP4, Anti-OATPRP4, Anti-Organic anion transporter polypeptide-related protein 4, Anti-Solute carrier family 21 member 15, Anti-Solute carrier organic anion transporter family member 5A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SVDAVSDDDVLKEKSNNSEQADKKVSSMGFGKDVRDLPRAAVRILSNM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLCO5A1(81796)

General description

The gene SLCO5A1 (solute carrier organic anion transporter family member 5A1) is mapped to human chromosome 8q13.3. The mRNA is expressed in the brain, heart, skeletal muscle and ovary. In addition, it is also expressed in bone tumors, prostate cancer, breast cancer, drug-resistant small cell lung cancer and liver cancer. The SLCO5A1 protein belongs to the OATP5 (organic anion-transporting polypeptide 5) family of proteins. The protein has 12 transmembrane domains and shows different localization depending on the tissue.

Immunogen

Solute carrier organic anion transporter family member 5A1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SLCO5A1 antibody produced in rabbit has been used in Western blotting, immunofluorescence and flow cytometry.

Biochem/physiol Actions

SLCO5A1 (solute carrier organic anion transporter family member 5A1) might be involved in cellular responses associated with the reorganization of the cell morphology, for instance differentiation and migration.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76253

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Olszewski-Hamilton et al.
Biomarkers in cancer, 3, 31-40 (2011-01-01)
The role of organic anion transporting polypeptide 5A1 (OATP5A1) a member of a family of drug transporters that mediate cellular uptake of drugs has not been characterized so far. Gene expression levels of OATP5A1 in small cell lung cancer (SCLC)
Izabel Patik et al.
Biochemical pharmacology, 98(4), 649-658 (2015-09-30)
Organic Anion Transporting Polypeptides (OATPs), encoded by genes of the Solute Carrier Organic Anion (SLCO) family, are transmembrane proteins involved in the uptake of various compounds of endogenous or exogenous origin. In addition to their physiological roles, OATPs influence the
Bertrand Isidor et al.
American journal of human genetics, 87(1), 95-100 (2010-07-07)
Mesomelia-synostoses syndrome (MSS) or mesomelic dysplasia with acral synostoses Verloes-David-Pfeiffer type is a rare autosomal-dominant disorder characterized by mesomelic limb shortening, acral synostoses, and multiple congenital malformations. So far, five patients in four unrelated families have been reported worldwide with
Katrin Sebastian et al.
PloS one, 8(12), e83257-e83257 (2014-01-01)
Organic anion transporting polypeptides (OATP/SLCO) have been identified to mediate the uptake of a broad range of mainly amphipathic molecules. Human OATP5A1 was found to be expressed in the epithelium of many cancerous and non-cancerous tissues throughout the body but

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service