Skip to Content
Merck
All Photos(6)

Documents

HPA019371

Sigma-Aldrich

Anti-ACO1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Aconitase, Anti-Citrate hydro-lyase, Anti-Cytoplasmic aconitate hydratase, Anti-IRE-BP 1, Anti-IRP1, Anti-Iron-responsive element-binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YERIHRSNLVGMGVIPLEYLPGENADALGLTGQERYTIIIPENLKPQMKVQVKLDTGKTFQAVMRFDTDVELTYFLNGGILNYMIRKMAK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACO1(48)

General description

The gene ACO1 (aconitase) is mapped to human chromosome 9p21.3-p21.1. It is also referred as IRBP (iron-responsive element-binding protein).

Immunogen

Cytoplasmic aconitate hydratase recombinant protein epitope signature tag (PrEST)

Application

Anti-ACO1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

ACO1 (aconitase) has dual roles. In iron-limiting conditions, it behaves as an RNA binding protein and controls the expression of genes involved in uptake, sequestration, and utilization of iron. In addition, it binds with a [4Fe-4S] cluster and functions as an enzyme which interconverts citrate and isocitrate. SIRT3 (NAD-dependent protein deacetylase sirtuin-3, mitochondrial) controls iron metabolism in cells and cancer growth by suppressing ACO1.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75234

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kurtis J Haro et al.
PloS one, 7(11), e48841-e48841 (2012-11-17)
Iron is required for nearly all organisms, playing important roles in oxygen transport and many enzymatic reactions. Excess iron, however, can be cytotoxic. Emerging evidence suggests that radioresistance can be achieved in lower organisms by the protection of proteins, but
Ji Un Kang et al.
International journal of oncology, 37(2), 327-335 (2010-07-03)
Chromosomal alterations are a major genomic force contributing to the development of lung cancer. We subjected 22 cases of squamous cell carcinoma of the lung (SCC) to whole-genome microarray-CGH (resolution, 1 Mb) to identify critical genetic landmarks that might be
Jean-Marc Blouin et al.
Blood, 136(21), 2457-2468 (2020-07-18)
Congenital erythropoietic porphyria (CEP) is an inborn error of heme synthesis resulting from uroporphyrinogen III synthase (UROS) deficiency and the accumulation of nonphysiological porphyrin isomer I metabolites. Clinical features are heterogeneous among patients with CEP but usually combine skin photosensitivity
Jérôme Dupuy et al.
Structure (London, England : 1993), 14(1), 129-139 (2006-01-13)
Iron regulatory proteins (IRPs) control the translation of proteins involved in iron uptake, storage and utilization by binding to specific noncoding sequences of the corresponding mRNAs known as iron-responsive elements (IREs). This strong interaction assures proper iron homeostasis in animal
C C Philpott et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(15), 7321-7325 (1994-07-19)
The iron-responsive element binding protein/cytosolic aconitase functions as either an RNA binding protein that regulates the uptake, sequestration, and utilization of iron or an enzyme that interconverts citrate and isocitrate. These mutually exclusive functions are regulated by changes in cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service